Order Entry
Germany
ContactUsLinkComponent
Mineral oil, white (petroleum) light, high purity
Irritant
Mineral oil, white (petroleum) light, high purity
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Mineral oil, white (petroleum) light, high purity
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Specific Gravity:
    0.838 – 0.854
  • Viscosity @ 40 °C:
    14.2 – 17 cS

Specification Test Results

Related Information

Frequently Bought Together