Order Entry
Germany
ContactUsLinkComponent
Coomassie® brilliant blue R-250 electrophoresis gel stain
Coomassie® brilliant blue R-250 electrophoresis gel stain
Catalog # 786-495
Supplier:  G-Biosciences
CAS Number:  
Coomassie® brilliant blue R-250 electrophoresis gel stain
Catalog # 786-495
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :825.98 g/mol
Melting Pt:174…180 °C
Storage Temperature:Ambient
EINECS:228-060-5
CAS Number:6104-59-2
Formula:C₄₅H₄₄N₃NaO₇S₂
MDL Number:MFCD00041762