Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250 electrophoresis gel stain
Coomassie brilliant blue G-250 electrophoresis gel stain
Coomassie brilliant blue G-250 electrophoresis gel stain
786-496
:
G-Biosciences
:
Coomassie brilliant blue G-250 electrophoresis gel stain
786-496
:
G-Biosciences
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
854.04 g/mol
:
Min, 1 g/cm³ (20 °C)
:
228-058-4
:
GHS07
:
Ambient
:
6104-58-1
:
C₄₇H₄₈N₃NaO₇S₂
:
MFCD00078482