Order Entry
Germany
ContactUsLinkComponent
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
Catalog # SIALN0786-15VL
Supplier:  Merck
CAS Number:  
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
Catalog # SIALN0786-15VL
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    15 Vial
  • Seq:
    0277

Specifications

About this item

 :
 :709.41 g/mol
EINECS:210-123-3
CAS Number:606-68-8
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number:MFCD00036200