Order Entry
Germany
ContactUsLinkComponent
Titanium standard solution, 1,000 mg/l Ti in water (from (NH4)2TiF6) standard for AAS and ICP
Titanium standard solution, 1,000 mg/l Ti in water (from (NH4)2TiF6) standard for AAS and ICP
Catalog # PERCN9300162
Supplier:  PerkinElmer
CAS Number:  
Titanium standard solution, 1,000 mg/l Ti in water (from (NH4)2TiF6) standard for AAS and ICP
Catalog # PERCN9300162
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O/tr 0,24% F-
  • Pk:
    500 mL
  • Element:
    Titanium
  • Symbol:
    Ti

Specifications

About this item