Order Entry
Germany
ContactUsLinkComponent
 
Molybdenum standard solution, 1000 mg/L Mo in water (from (NH₄)₆Mo₇O₂₄) standard for AAS and ICP
  PERCN9300134
 :  
Molybdenum standard solution, 1000 mg/L Mo in water (from (NH₄)₆Mo₇O₂₄) standard for AAS and ICP
  PERCN9300134
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    1
  • Element:
    Molybdenum
  • Symbol:
    Mo