β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form), Sigma-Aldrich®
Catalog # SIALN8535-15VL
Supplier: Merck
CAS Number:
Specifications
- Pk:15 Vial
- Seq:0277
Specifications
About this item
:
:663.43 g/mol
Merck Index:13,06370
Melting Pt:140 to 142 °C
Storage Temperature:Refrigerator
EINECS:200-184-4
CAS Number:53-84-9
Formula:C₂₁H₂₇N₇O₁₄P₂
MDL Number:MFCD00036253