Order Entry
Germany
ContactUsLinkComponent
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form), Sigma-Aldrich®
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form), Sigma-Aldrich®
Catalog # SIALN8410-15VL
Supplier:  Merck
CAS Number:  
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form), Sigma-Aldrich®
Catalog # SIALN8410-15VL
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    15 Vial
  • Seq:
    0277

Specifications

About this item

 :
 :663.43 g/mol
Merck Index:13,06370
Melting Pt:140 to 142 °C
Storage Temperature:Refrigerator
EINECS:200-184-4
CAS Number:53-84-9
Formula:C₂₁H₂₇N₇O₁₄P₂
MDL Number:MFCD00036253