Orders LinkContactUsLinkComponent
Adapter, stopper type, DURAN®
Adapter, stopper type, DURAN®
Supplier:   DWK Life Sciences
Adapter, stopper type, DURAN®
Supplier:  DWK Life Sciences
Certificates
Adapter, stopper type, DURAN®
Adapter, stopper type, DURAN®
6 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, stopper type, solid, socket and cone.
Product Family Options
Product Information
  • Type
    Cone size
    Socket size
    Availability
    Price
  • stopper, solid
    29/32 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 391,00 €
    391,00 €
    1 * 10 ST
       391,00 €
  • stopper, hollow
    29/32 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 362,00 €
    362,00 €
    1 * 10 ST
       362,00 €
  • stopper, solid
    19/26 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 348,00 €
    348,00 €
    1 * 10 ST
       348,00 €
  • stopper, solid
    24/29 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 362,00 €
    362,00 €
    1 * 10 ST
       362,00 €
  • stopper, solid
    24/29 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 362,00 €
    362,00 €
    1 * 10 ST
       362,00 €
  • stopper, solid
    29/32 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 390,00 €
    390,00 €
    1 * 10 ST
       390,00 €