Petri dishes, coloured, Sterilin®
Supplier: Sterilin
Certificates
About this item
PS, with lid. Colour coded Petri dishes especially suitable for the identification of group work within teaching laboratories.
- Aids ease of identification
- Aseptically manufactured under cleanroom conditions (class 7 ISO14644) to exclude microbiological contamination
- Produced and tested in accordance with the BS611 part 2 standard
- Manufactured using cadmium free non cytotoxic colourants
Aseptically manufactured, In vitro use only. Perform non standard applications with Thermo Scientific™ Sterilin™ special Petri dishes. minimum order quantities apply. Not available in America.