Extraction thimbles, high purity, glass, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
Cytiva's Whatman 100% borosilicate glass microfiber (HP-GF) thimbles are designed to fit most Soxhlet extraction units. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.
- Resistant to temperatures up to 500 °C
- Binder-free, no chemical additives
- Designed to fit most commercially available Soxhlet extractors
- Effective when used with solvents incompatible with cellulose materials
Glass Soxhlet extraction thimble applications:
Analysis of hot and acidic gases during emission control and smoke stack gas monitoring
Analysis of pesticide residues using Soxhlet extraction
Determination of oil/fat content of foods
Analysis of oil and grease in solid wastes
Whatman brand high-purity glass thimbles are available in a range of sizes and wall thicknesses for optimal use in a variety of air sampling and Soxhlet extraction units.