ContactUsLinkComponent
Beaker tongs, stainless steel
Beaker tongs, stainless steel
  NICK7005
undefined
Beaker tongs, stainless steel
  NICK7005
  • Length:
    356 mm
  • Pk:
    1
These stainless steel beaker tongs feature silicone covered jaws.
  • Silicone covered sleeves reduce risk breakage when handling beakers
  • Operating temperature: −60...+180˚C
  • Hold beakers from 100 to 1000 ml