Flash chromatography columns, PuriFlash®, C18 AQ silica
These flash chromatography columns, C18 AQ silica are suitable for the purification of mid polar compounds.
- 100% water compatible
- Base material is high performance spherical silica
- pH stability ranges from 1,5 to 7,5