PIG® BLUE absorbent socks
Supplier: NEW PIG
Certificates
About this item
Surround spills to prevent spreading. Extra-fine vermiculite filler pulls in liquid and hugs floors to stay in place.
- Heaviest sock, features extra-dense construction to hug floors and stay in place whilst containing spills
- Fine-grade vermiculite filler quickly absorbs leaks, drips and spills; great for spill response or absorbing machine leaks
- Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated
- Absorbs and retains oils, coolants, solvents and water
Ideal for diking and damming liquids. Can be air-dried for reuse when soaked with water.