Absorbent booms, oil-only, Versatile PIG®
Supplier: NEW PIG
Certificates
About this item
Booms made from PP skin (UV resistant up to 12 months), PP filler and a tough outer mesh which is UV resistant for long-term outdoor use without degradation. The PP filler is chemical-resistant and hydrophobic for containing and absorbing oil-based spills. The booms float at water surface for easy retrieval, even when saturated.
- Rope running along length of boom withstands up to 400-lb. loads, adding strength for deployment and retrieval
- Strong, zinc-plated carbon steel attachment clips and rings allow you to link booms together for greater length
- Bright white colour makes booms easier to see in outdoor environments and clearly shows saturation level
- Can be incinerated after use to reduce waste or for fuels blending
Field-proven performance works in all conditions to absorb oil and oil-based liquids, including lubricants and fuel, on land or water.