ContactUsLinkComponent
Crucible, quartz
Crucible, quartz
Catalog # 459-0181
Crucible, quartz
Catalog # 459-0181
Specifications
  • Ø×H:
    25×38 mm
  • Type:
    with projections
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
This crucible and lid are made of quartz glass.
Lid
Product Information