Orders LinkContactUsLinkComponent
Hydrometers
Hydrometers
Supplier:   Alla France
Hydrometers
Supplier:  Alla France
Certificates
Hydrometers
10 Options Available Below
Product Details & Documents
Handelsübliche Dichte-Aräometer mit einer Bezugstemperatur von 20 °C.
Product Family Options
Product Information
  • Messbereich
    Teilung
    Länge
    Availability
    Price
  • 2,000 - 2,200 g/ml
    0,002 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 265,00 €
    265,00 €
    1 * 1 ST
       265,00 €
  • 2,200 - 2,400 g/ml
    0,002 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 384,00 €
    384,00 €
    1 * 1 ST
       384,00 €
  • 2,400 - 2,600 g/ml
    0,002 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 384,00 €
    384,00 €
    1 * 1 ST
       384,00 €
  • 2,600 - 2,800 g/ml
    0,002 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 384,00 €
    384,00 €
    1 * 1 ST
       384,00 €
  • 2,800 - 3,000 g/ml
    0,002 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 265,00 €
    265,00 €
    1 * 1 ST
       265,00 €
  • 2,000 - 2,500 g/ml
    0,005 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 455,00 €
    455,00 €
    1 * 1 ST
       455,00 €
  • 2,500 - 3,000 g/ml
    0,005 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 455,00 €
    455,00 €
    1 * 1 ST
       455,00 €
  • 3,000 - 3,500 g/ml
    0,005 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 455,00 €
    455,00 €
    1 * 1 ST
       455,00 €
  • 2,000 - 3,000 g/ml
    0,010 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 682,00 €
    682,00 €
    1 * 1 ST
       682,00 €
  • 3,000 - 4,000 g/ml
    0,010 g/ml
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 682,00 €
    682,00 €
    1 * 1 ST
       682,00 €