Safety transport containers
Supplier: ROETZMEIER SICHERH BEHAELTER
Certificates
About this item
Safety transport containers for UN approved transport of hazardous goods on public routes.
- High quality, and chemically resistant stainless steel with PTFE gaskets
- All types of hazardous goods (UN X approval)
- Help to avoid non returnable one-way transport packaging
- Screw cap without pressure control valve - also available with pressure control valve as well
- Removable flame trap
Design approved according to ADR, RID, IMDG Code, ICAO-TI and GGVSEB.
Certifications: Design approved according to ADR, RID, IMDG Code, ICAO-TI and GGVSEB.
Delivery: 10 and 25 litre barrels, and 20 litre canisters are equipped with separate ventilation screw to maximise the flow rate and allow for decanting in horizontal position, when used in conjunction with self-closing tap (119-2401).