Flasks, round bottom, with standard ground joints
Supplier: witeg Labortechnik
Certificates
About this item
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.