Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Deoxyuridine
2'-Deoxyuridine, Sigma-Aldrich®
2'-Deoxyuridine, Sigma-Aldrich®
2'-Deoxyuridine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2'-Deoxyuridine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
951-78-0
:
Formula:
C₉H₁₂N₂O₅
MDL Number:
MFCD00006527
:
228.20 g/mol
:
M-Clarity™ quality level = MQ100
Storage Temperature:
Ambient
EINECS:
213-455-7