AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
CAS Number:30827-99-7
:
Formula:C₈H₁₁ClFNO₂S
MDL Number:MFCD00132962
:239.69 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS07