N,N'-Di-sec-butyl-p-phenylenediamine
Catalog # ABCRAB141113.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
Specifications
About this item
:
:220.3500 g/mol
EINECS:202-992-2
CAS Number:101-96-2
Formula:C₁₄H₂₄N₂
MDL Number:MFCD00043658