Bovine alpha-Chymotrypsin type I-S (from Pancreas)
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:9004-07-3
- Source:Pancreas
- Species:Bovine
- Storage conditions:Freezer
- MDL No.:MFCD00130481
- Enzyme name:alpha-Chymotrypsin type I-S
Specifications
Frequently Bought Together