About this item
Chymotrypsin (Chy) is a serine protease. It corresponds to a molecular weight of 25,7 kDa and is widely used in pharmaceutical industry.It is synthesized in pancreas from chymotrypsinogen and require calcium for this conversion.α-Chymotrypsin from bovine pancreas (bovine pancreatic α-chymotrypsin, CHT) is an enzyme protein.The influence of varying concentrations of organic solvents like ethanol, 1,4-dioxane and acetonitrile on CHT has been reported.
α-Chymotrypsin from bovine pancreas is used for the protein Identification by mass spectrometry (MS), the isolation and characterization of myosin heavy chains, toxin preparation, the incubation of infected and uninfected cells for analysis of cellular proteins by SDS-PAGE.α-Chymotrypsin from bovine pancreas (BPC) may be used as a catalyst in the preparation of tetrahydroquinoline derivatives by Povarov reaction.
Specifications
- CAS No.:9004-07-3
- Storage conditions:Freezer
- Form:Lyophilised powder
- MDL No.:MFCD00130481
- Enzyme name:alpha-Chymotrypsin