Order Entry
Germany
ContactUsLinkComponent
alpha-Chymotrypsin
alpha-Chymotrypsin
Supplier:  Merck
Certificates
alpha-Chymotrypsin
Supplier:  Merck

Specifications

  • CAS No.:
    9004-07-3
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00130481
  • Enzyme name:
    alpha-Chymotrypsin
  • Enzyme activity:
    ≥45 U/mg

Specifications