Order Entry
Germany
ContactUsLinkComponent
 
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)
Catalog # MOLEM73580995
Supplier:  Molekula
CAS Number:  
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)
Catalog # MOLEM73580995
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :363.5988 g/mol
Storage Temperature:Ambient
EINECS:239-003-9
CAS Number:14933-09-6
Formula:C₁₉H₄₁NO₃S
MDL Number:MFCD00036910