N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14), Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:14933-09-6
:
Formula:C₁₉H₄₁NO₃S
MDL Number:MFCD00036910
:363.5988 g/mol
Storage Temperature:Ambient
EINECS:239-003-9