Order Entry
Germany
ContactUsLinkComponent
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
Corrosive
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
Catalog # MOLEM74706973
Supplier:  Molekula
CAS Number:  
Corrosive
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
Catalog # MOLEM74706973
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :335.55 g/mol
Storage Temperature:Ambient
EINECS:239-002-3
UN:3261
ADR:8,II
 :GHS05
CAS Number:14933-08-5
Formula:C₁₇H₃₇NO₃S
MDL Number:MFCD00036909