Order Entry
Germany
ContactUsLinkComponent
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10)
Irritant
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10)
Catalog # MOLE20889868-25G
Supplier:  Molekula
CAS Number:  
Irritant
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10)
Catalog # MOLE20889868-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :307.4924 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:15163-36-7
Formula:C₁₅H₃₃NO₃S
MDL Number:MFCD00036908