Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Betaine hydrochloride
Betaine hydrochloride, Sigma-Aldrich®
Betaine hydrochloride, Sigma-Aldrich®
Betaine hydrochloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Betaine hydrochloride, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
590-46-5
:
Formula:
C₅H₁₂ClNO₂
MDL Number:
MFCD00011903
:
153.61 g/mol
:
M-Clarity™ quality level = MQ100
Storage Temperature:
Ambient
EINECS:
209-683-1