Betaine hydrochloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:590-46-5
:
Formula:C₅H₁₂ClNO₂
MDL Number:MFCD00011903
:153.61 g/mol
:M-Clarity™ quality level = MQ200
EINECS:209-683-1
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:647 g/l (25 °C)
- Melting Point:242 - 246 °C
- Molar Mass:153.61 g/mol
- pH:1 (50 g/l, H2O, 20 °C)
- Ignition Point:400 °C