Betaine hydrochloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:590-46-5
:
Formula:C₅H₁₂ClNO₂
MDL Number:MFCD00011903
:153.61 g/mol
Merck Index:14,1179
Melting Pt:∼ 240 °C (dec.)
EINECS:209-683-1
:GHS07
Storage Temperature:Ambient