Lithium chloride 1 mol/l in ethanol, Supelco®
Catalog # 62481-250ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:42.39 g/mol
Storage Temperature:Ambient
EINECS:231-212-3
UN:1170
ADR:3,II
:GHS02
CAS Number:7447-41-8
Formula:LiCl
MDL Number:MFCD00011078