Sodium carbonate 0.025 mol/l (0.05 N) in aqueous solution volumetric solution
Catalog # CHMP818095811.1000
Supplier: CHEMPUR
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:105.99 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494