Cut resistant gloves, PU800
This seamless polyethylene High Tenacity/polyester knitted liner features a PU coating on the palm. It provides excellent abrasion and blade cuts resistance.
- Fine and stretchable stitches for excellent comfort and adaptability
- Latex knit wrist to ensure snug fit
- High flexibility and dexterity for an optimum comfort in use
Ideal for fine tactile handling, sharp components handling, fitting and assembling works, quality control, automotive industry and glass industry.
EN 388 (4342), Cat. II
: EN 388 (4342), Cat. II
- Catalog No:
- MARG1872E04000
- MARG1872E02000
- MARG1872E03000
- MARG1872E01000
- Model:
- PU800
- PU800
- PU800
- PU800
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyester
- Polyester
- Polyester
- Polyester
- Finish:
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (4342)
- EN 388 (4342)
- EN 388 (4342)
- EN 388 (4342)