ContactUsLinkComponent
Flame retardant rain jacket, Hasnon 3085
 :  SIOEN
undefined
Flame retardant rain jacket, Hasnon 3085
 :  SIOEN
  • Catalog No:
  • 113-7087
  • 113-7086
  • 113-7089
  • 113-7090
  • 113-7091
  • 113-7088
  • Model:
  • 3085 Hasnon Siopor
  • 3085 Hasnon Siopor
  • 3085 Hasnon Siopor
  • 3085 Hasnon Siopor
  • 3085 Hasnon Siopor
  • 3085 Hasnon Siopor
  • Garment application:
  • Fire protection
  • Fire protection
  • Fire protection
  • Fire protection
  • Fire protection
  • Fire protection
  • Risk area:
  • Rain (waterproof), Flame-retardant
  • Rain (waterproof), Flame-retardant
  • Rain (waterproof), Flame-retardant
  • Rain (waterproof), Flame-retardant
  • Rain (waterproof), Flame-retardant
  • Rain (waterproof), Flame-retardant
  • Gender:
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • 100% polyester with 100% FR PU coating + AST
  • 100% polyester with 100% FR PU coating + AST
  • 100% polyester with 100% FR PU coating + AST
  • 100% polyester with 100% FR PU coating + AST
  • 100% polyester with 100% FR PU coating + AST
  • 100% polyester with 100% FR PU coating + AST
  • Weight:
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²