Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Head Protection
Safety Helmets, customised
Safety helmet with logo MA07, concept
Safety helmet with logo MA07, concept
Safety helmet with logo MA07, concept
CENUS09EWF/M.LOGO
:
CENTURION SAFETY PRODUCTS
Safety helmet with logo MA07, concept
CENUS09EWF/M.LOGO
:
CENTURION SAFETY PRODUCTS
Colour:
White
Description:
Hard hat, Centurion™ Concept, with "MA07" logo
Pk:
1