Orders LinkContactUsLinkComponent
Harnesses, Titan™
Harnesses, Titan™
Harnesses, Titan™
Harnesses, Titan™
Non stretch harnesses for construction work. Available as a 1-point version with rear anchorage or as a 2-point version with front and rear anchorage.
  • Lightweight, durable polyester webbing for all-day comfort and dependability
  • Fully adjustable designs so one size fits all
  • Sliding back D-ring for a comfortable fit
  • Sub pelvic strap keeps the wearer upright
  • Chest strap (45 mm) with metal buckles
EN 361
 : EN 361
Product Family Options
  • Description
    Weight
    Attachment Points
  • Harness, Polyamide
    0,6 kg
    2-point
    1 * 1 ST
    1 * 1 ST - 85,90 €
    85,90 €
    1 * 1 ST
       85,90 €
  • Harness, Polyamide
    0,5 kg
    1 * 1 ST
    1 * 1 ST - 63,90 €
    63,90 €
    1 * 1 ST
       63,90 €