Fall protection kit, Miller® Titan™
Catalog # 139-0234
Supplier: HONEYWELL SAFETY
Specifications
- Description:Fall protection kit for construction workers
- Length:2 m
- Pk:1
Specifications
About this item
Safety kit for maintenance work at industrial plants, tasks on antennae and construction sites.
Certifications: EN 361, EN 355, EN 354.
Delivery: The kit contains:
Titan™ single point safety harness with safety loop at the back
Lanyard, 2 m, with shock absorber
Two screwgate karabiners
Titan™ single point safety harness with safety loop at the back
Lanyard, 2 m, with shock absorber
Two screwgate karabiners