Glove box holders, Poxygrid®, SP Bel-Art
Epoxy-coated, welded steel wire racks for one, two or three boxes of disposable gloves, wipes or tissues.
- Glove box drops into an individual retaining compartment and lifts out easily for replacement
- Designed for mounting to a vertical surface
- Durable, blue epoxy coated racks are steam autoclavable at 121 °C