ContactUsLinkComponent
Bags, safety pouch, SP Bel-Art
Bags, safety pouch, SP Bel-Art
  BELAH132340000
undefined
Bags, safety pouch, SP Bel-Art
  BELAH132340000
  • Packed:
    200 pcs/pack
  • Description:
    Biohazard sharp object safety pouch, thickness 0,25 mm
  • L×W:
    140×330 mm
  • Material:
    Paperboard
  • Pk:
    200
Hard to pierce pouch offering protection against the danger of being cut by contaminated sharp objects. Convenient for labs, clinics and medical offices. Used with the Poxygrid® safety pouch stand for easy benchtop collection.
  • Solid paperboard pouch
  • Remains open in an oval shape by folding the bottom flaps
  • After filling, the top flaps close and lock in place for safe disposal
  • Printed with bold biohazard symbol