Orders LinkContactUsLinkComponent
Double spatulas, powder
Double spatulas, powder
Supplier:   RSG
Double spatulas, powder
Supplier:  RSG
Certificates
Double spatulas, powder
Double spatulas, powder
4 Options Available Below
Product Details & Documents
Stainless steel.
  • With round handle, one flat and one scoop shaped, rounded spatula end
  • Straight shape
  • Very good thermal resistance, autoclavable
  • Good chemical resistance
Product Family Options
Product Information
  • Length
    Blade width
    Availability
    Price
  • 185 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,10 €
    9,10 €
    1 * 1 ST
       9,10 €
  • 130 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 42,60 €
    42,60 €
    1 * 5 ST
       42,60 €
  • 150 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 43,90 €
    43,90 €
    1 * 5 ST
       43,90 €
  • 210 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 45,90 €
    45,90 €
    1 * 5 ST
       45,90 €