Order Entry
Germany
ContactUsLinkComponent
Tetracycline hydrochloride, Sigma-Aldrich®
Tetracycline hydrochloride, Sigma-Aldrich®
Catalog # SIALT4062-5G
Supplier:  Merck
CAS Number:  
Tetracycline hydrochloride, Sigma-Aldrich®
Catalog # SIALT4062-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :480.9 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:200-593-8
 :GHS07
Storage Temperature:Ambient
CAS Number:64-75-5
Formula:C₂₂H₂₅ClN₂O₈
MDL Number:MFCD00078142