Order Entry
Germany
ContactUsLinkComponent
Chlortetracycline hydrochloride can be used as secondary standard
Chlortetracycline hydrochloride can be used as secondary standard
Certificates
undefined
Chlortetracycline hydrochloride can be used as secondary standard

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Purity:
    ≥80 % (as is, calculated)

Specification Test Results

Related Information