Order Entry
Germany
ContactUsLinkComponent
Buffer, TRIS-tricine-SDS solution 10X concentrate, Sigma-Aldrich®
Buffer, TRIS-tricine-SDS solution 10X concentrate, Sigma-Aldrich®
Catalog # SIALT1165-500ML
Supplier:  Merck
CAS Number:  
Buffer, TRIS-tricine-SDS solution 10X concentrate, Sigma-Aldrich®
Catalog # SIALT1165-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    TRIS-tricine-SDS buffer solution 10X concentrate
  • M-Clarity quality level:
    MQ200

Specifications

About this item