Order Entry
Germany
ContactUsLinkComponent
Buffer, TRIS-glycine solution 10X concentrate (TG buffer), Sigma-Aldrich®
Buffer, TRIS-glycine solution 10X concentrate (TG buffer), Sigma-Aldrich®
Catalog # SIALT4904-1L
Supplier:  Merck
CAS Number:  
Buffer, TRIS-glycine solution 10X concentrate (TG buffer), Sigma-Aldrich®
Catalog # SIALT4904-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TRIS-glycine buffer solution 10X concentrate (TG buffer)

Specifications

About this item