Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer), Sigma-Aldrich®

Buffer, TE solution 100x concentrate (TRIS-EDTA buffer), Sigma-Aldrich®

TE buffer solution 100x concentrate (TRIS-EDTA buffer), Glass bottle, 100 mL
Artikelnummer SIALT9285-100ML
Hersteller:   Merck
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer), Sigma-Aldrich®
Artikelnummer SIALT9285-100ML
Hersteller:  Merck
CAS-Nummer:  
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer), Sigma-Aldrich®

TE buffer solution 100x concentrate (TRIS-EDTA buffer), Glass bottle, 100 mL

CAS-Nummer:  
Preis
55,40 €
Log in for availability
1 * 100 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    TE buffer solution 100x concentrate (TRIS-EDTA buffer)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.