Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer, TE solution 100x concentrate (TRIS-EDTA buffer)

TE buffer solution 100x concentrate (TRIS-EDTA buffer), Glass bottle, 1 L
Catalog # SERA42554.01
Marketsource
Supplier:   Serva
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer)
Catalog # SERA42554.01
Supplier:  Serva
Marketsource
CAS Number:  
Buffer, TE solution 100x concentrate (TRIS-EDTA buffer)

TE buffer solution 100x concentrate (TRIS-EDTA buffer), Glass bottle, 1 L

CAS Number:  
Price
61,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TE buffer solution 100x concentrate (TRIS-EDTA buffer)

Specifications