Order Entry
Germany
ContactUsLinkComponent
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # UPTIUP586046
Supplier:  Uptima
CAS Number:  
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # UPTIUP586046
Supplier:  Uptima
CAS Number:  

Specifications

  • Pk:
    4 L
  • Description:
    SSC buffer solution 20X concentrate (saline-sodium citrate)

Specifications

About this item