Ampicillin trihydrate, Sigma-Aldrich®
Catalog # SIALA1593-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:403.45 g/mol
:M-Clarity™ quality level = MQ200
EINECS:200-709-7
:GHS07
Storage Temperature:Refrigerator
CAS Number:7177-48-2
Formula:C₁₆H₁₉N₃O₄S·3H₂O
MDL Number:MFCD00072036